Ethyl-3-(2-furyl)-1H-pyrazole-5-carboxylate 98%

Lieferant: Thermo Scientific

Synonyme: Ethyl 3-(furan-2-yl)-1H-pyrazole-5-carboxylate

H25791.06
H25791.06EA 104 EUR
H25791.06
Ethyl-3-(2-furyl)-1H-pyrazole-5-carboxylate 98%
Ethyl-3-(2-furyl)-1H-pyrazole-5-carboxylate

Purity: 98% CAS No.: 33545-44-7 Molecular Formula: C10H10N2O3 Formula Weight: 206.20 Melting Point: 135-137° MDL No.: MFCD09953454

Formel: C₁₀H₁₀N₂O₃
Molecular Weight: 206,20 g/mol
Schmelzpunkt: 135…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09953454
CAS-Nummer: 33545-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR