5-Brom-4,6-dichlorpyrimidin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
H25821.03
H25821.03EA
110
EUR
H25821.03
5-Brom-4,6-dichlorpyrimidin ≥97%
5-Bromo-4,6-dichloropyrimidine
Purity: 97%
CAS No.: 68797-61-5
Molecular Formula: C4HBrCl2N2
Formula Weight: 227.88
Melting Point: 77-81°
UN No.: 1759
MDL No.: MFCD00047379
Formel:
C₄HBrCl₂N₂ Molecular Weight: 227,88 g/mol Schmelzpunkt: 77…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047379 CAS-Nummer: 68797-61-5 EINECS: 272-305-9 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...