6-Chlor-2-fluorpurin ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Chloro-2-fluoro-9H-purine
H25825.ME
H25825.MEEA
92
EUR
H25825.ME
6-Chlor-2-fluorpurin ≥97%
6-Chloro-2-fluoropurine
Purity: 97%
CAS No.: 1651-29-2
Molecular Formula: C5H2ClFN4
Formula Weight: 172.55
Melting Point: 151-153°
MDL No.: MFCD02183557
Formel:
C₅H₂ClFN₄ Molecular Weight: 172,55 g/mol Schmelzpunkt: 151…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02183557 CAS-Nummer: 1651-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...