4,4-Difluorcyclohexancarbonsäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4,4-difluorocyclohexane-1-carboxylic acid

H25831.03
H25831.03EA 134 EUR
H25831.03
4,4-Difluorcyclohexancarbonsäure 98%
4,4-Difluorocyclohexanecarboxylic acid

Purity: 98% CAS No.: 122665-97-8 Molecular Formula: C7H10F2O2 Formula Weight: 164.15 Melting Point: 104-107° MDL No.: MFCD03788493

Formel: C₇H₁₀F₂O₂
Molecular Weight: 164,15 g/mol
Schmelzpunkt: 104…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788493
CAS-Nummer: 122665-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR