2-(Trifluoromethyl)pyrrolidine 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-Aza-2-(trifluoromethyl)cyclopentane, 2-(Trifluoromethyl)tetrahydropyrrole

H25852.MD H25852.03
H25852.MDEA 77.5 EUR
H25852.MD H25852.03
2-(Trifluoromethyl)pyrrolidine 95%
2-(Trifluoromethyl)pyrrolidine

Purity: 95% CAS No.: 109074-67-1 Molecular Formula: C5H8F3N Formula Weight: 139.12 Flash Point: 26°(78°F) Refractive Index: 1.379 Boiling Point: 110-112° UN No.: UN2733 MDL No.: MFCD02663405

Formel: C₅H₈F₃N
Molecular Weight: 139.12 g/mol
Siedepunkt: 110…112 °C
Dichte: 1.21
Flammpunkt: 26 °C (78 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD02663405
CAS-Nummer: 109074-67-1
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR