(S)-2-(Trifluoromethyl)pyrrolidine 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H25854.MD
H25854.03
H25854.MDEA
37.3
EUR
H25854.MD
H25854.03
(S)-2-(Trifluoromethyl)pyrrolidine 95%
(S)-2-(Trifluoromethyl)pyrrolidine
Purity: 95%
CAS No.: 119580-41-5
Molecular Formula: C5H8F3N
Formula Weight: 139.12
Flash Point: 26°(79°F)
Melting Point: 27-31°
Boiling Point: 110-112°
Optical Rotation: +10° (c=1 in methanol)
UN No.: UN2925
MDL No.: MFCD03093766
Formel:
C₅H₈F₃N Siedepunkt: 110…112 °C Schmelzpunkt: 27…31 °C Flammpunkt: 26 °C (79 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093766 CAS-Nummer: 119580-41-5 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...