4,5,6-Trifluorpyrimidin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
H25855.03
H25855.03EA
177
EUR
H25855.03
4,5,6-Trifluorpyrimidin ≥97%
4,5,6-Trifluoropyrimidine
Purity: 97%
CAS No.: 17573-78-3
Molecular Formula: C4HF3N2
Formula Weight: 134.06
Flash Point:
Formel:
C₄HF₃N₂ Molecular Weight: 134,06 g/mol Siedepunkt: 83…85 °C Dichte: 1,4609 Flammpunkt: >85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD05662363 CAS-Nummer: 17573-78-3 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...