3,3,3-Trifluoropropane-1-sulphonyl chloride 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3,3,3-Trifluoropropane-1-sulphonylchloride

H25856.ME
H25856.MEEA 344 EUR
H25856.ME
3,3,3-Trifluoropropane-1-sulphonyl chloride 95%
3,3,3-Trifluoropropane-1-sulphonyl chloride

Purity: 95% CAS No.: 845866-80-0 Molecular Formula: C3F3H4ClO2S Formula Weight: 196.57 Refractive Index: 1.3997 Boiling Point: 77-78°/16mm UN No.: UN3265 MDL No.: MFCD06248898

Formel: C₃H₄ClF₃O₂S
Molecular Weight: 196,58 g/mol
Siedepunkt: 77…78 °C (16 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD06248898
CAS-Nummer: 845866-80-0
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR