4,4-Dimethylcyclohexanon ≥98%

Lieferant: Thermo Scientific

Synonyme: 4,4-Dimethylcyclohexan-1-one

H25857.03
H25857.03EA 83.8 EUR
H25857.03
4,4-Dimethylcyclohexanon ≥98%
4,4-Dimethylcyclohexanone

Purity: 98% CAS No.: 4255-62-3 Molecular Formula: C8H14O Formula Weight: 126.20 Flash Point: 62°(144°F) Melting Point: 41-43° MDL No.: MFCD00234965

Formel: C₈H₁₄O
Molecular Weight: 126,2 g/mol
Schmelzpunkt: 41…43 °C
Flammpunkt: 62 °C (144 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00234965
CAS-Nummer: 4255-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR