2-(Ethoxycarbonyl)phenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(Ethoxycarbonyl)benzeneboronic acid, (2-(Ethoxycarbonyl)phenyl)boronic acid

H25867.06 H25867.03
H25867.06EA 98.8 EUR
H25867.06 H25867.03
2-(Ethoxycarbonyl)phenylboronsäure ≥98%
2-(Ethoxycarbonyl)phenylboronsäure

Purity: 98% CAS No.: 380430-53-5 Molecular Formula: C9H11BO4 Formula Weight: 193.99 Melting Point: 130-133° MDL No.: MFCD02179453

Formel: C₉H₁₁BO₄
Schmelzpunkt: 130…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179453
CAS-Nummer: 380430-53-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR