Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine 94%

Lieferant: Thermo Fisher Scientific
Gefahr

H25882.06
H25882.06EA 638 EUR
H25882.06
Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine 94%
Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine

Purity: 94% CAS No.: 212713-08-1 Molecular Formula: C30H46ClO2P Formula Weight: 505.13 UN No.: UN3261 MDL No.: MFCD08458881

Formel: C₃₀H₄₆ClO₂P
Lagertemperatur: Raumtemperatur
MDL: MFCD08458881
CAS-Nummer: 212713-08-1
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR