Chlorbis(4-methoxyphenyl)phosphin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
H25887.03
H25887.03EA
123
EUR
H25887.03
Chlorbis(4-methoxyphenyl)phosphin ≥98%
Chlorbis(4-methoxyphenyl)phosphin
Purity: 98+%
CAS No.: 13685-30-8
Molecular Formula: C14H14ClO2P
Formula Weight: 280.69
Odour: Irritating
Melting Point: 61-64°
Boiling Point: 140°/1.5mm
UN No.: 3261
MDL No.: MFCD04972301
Formel:
C₁₄H₁₄ClO₂P Molecular Weight: 280,69 g/mol Siedepunkt: 140 °C (1,5 mmHg) Schmelzpunkt: 61…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972301 CAS-Nummer: 13685-30-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...