Chlorbis(2-methylphenyl)phosphin 98%, Stäbe
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Di-o-tolylchlorophosphine, Chlorodi(o-tolyl)phosphine
H25890.06
H25890.06EA
265
EUR
H25890.06
Chlorbis(2-methylphenyl)phosphin 98%, Stäbe
Chlorbis(2-methylphenyl)phosphin
Purity: 98%
CAS No.: 36042-94-1
Molecular Formula: C14H14ClP
Formula Weight: 248.69
Melting Point: 57°
Boiling Point: 174-178°/3mm
UN No.: 1760
MDL No.: MFCD04038733
Formel:
C₁₄H₁₄ClP Molecular Weight: 248.688 g/mol Siedepunkt: 174…178 °C (3 mmHg) Schmelzpunkt: 57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038733 CAS-Nummer: 36042-94-1 UN: 1760 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...