(E)-3,4-Dihydroxybenzylideneacetone 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H25901.06
H25901.06EA
275
EUR
H25901.06
(E)-3,4-Dihydroxybenzylideneacetone 97%
(E)-3,4-Dihydroxybenzylideneacetone
Purity: 97%
CAS No.: 123694-03-1
Molecular Formula: C10H10O3
Formula Weight: 178.19
Melting Point: 173-175°
MDL No.: MFCD00916691
Formel:
C₁₀H₁₀O₃ Schmelzpunkt: 173…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00916691 CAS-Nummer: 123694-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...