(R)-(+)-2-(Boc-Aminomethyl)-4,4-diethoxypiperidine fumarate

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2R)-2-(Aminomethyl)-4,4-diethoxypiperidine fumarate, 2-BOC protected

H25909.ME
H25909.MEEA 133 EUR
H25909.ME
(R)-(+)-2-(Boc-Aminomethyl)-4,4-diethoxypiperidine fumarate
(R)-(+)-2-(Boc-Aminomethyl)-4,4-diethoxypiperidine fumarate

Molecular Formula: C15H30N2O4.C4H4O4 Formula Weight: 418.49 MDL No.: MFCD09953465

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR