Albendazole sulphone

Lieferant: Thermo Scientific
Achtung

H25917.03
H25917.03EA 248 EUR
H25917.03
Albendazole sulphone
Albendazole sulphone

CAS No.: 75184-71-3 Molecular Formula: C12H15N3O4S Formula Weight: 297.33 Melting Point: 290-292° MDL No.: MFCD00600775

Formel: C₁₂H₁₅N₃O₄S
Molecular Weight: 297,33 g/mol
Schmelzpunkt: 290…292 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00600775
CAS-Nummer: 75184-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR