2,7-Dibrom-9-fluorenon 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,7-Dibromo-9H-fluoren-9-one

H25919.06 H25919.03
H25919.06EA 98.6 EUR
H25919.06 H25919.03
2,7-Dibrom-9-fluorenon 96%
2,7-Dibrom-9-fluorenon

Purity: 96% CAS No.: 14348-75-5 Molecular Formula: C13H6Br2O Formula Weight: 337.99 Melting Point: 195-196° MDL No.: MFCD00010790

Formel: C₁₃H₆Br₂O
Molecular Weight: 338 g/mol
Schmelzpunkt: 195…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010790
CAS-Nummer: 14348-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR