D-Phenylalanin-methylester Hydrochlorid 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (R)-Methyl 2-amino-3-phenylpropanoate hydrochloride, H-D-Phe-OMe.HCl, H-D-Phe-OMe·HCl

H25920.14 H25920.06
H25920.14EA 150 EUR
H25920.14 H25920.06
D-Phenylalanin-methylester Hydrochlorid 99%
D-Phenylalanine methyl ester hydrochloride

Purity: 99% CAS No.: 13033-84-6 Molecular Formula: C10H13NO2.HCl Formula Weight: 215.68 Melting Point: 158-162° MDL No.: MFCD00066112

Formel: C₁₀H₁₃NO₂
Schmelzpunkt: 158…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066112
CAS-Nummer: 13033-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR