Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl ethoxymethylene-3-oxo-4,4,4-trifluorobutyrate, 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylic Acid Ethyl Ester, Ethyl 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylate, 2-(Ethoxymethylene)-4,4,4-trifluoro-3-oxobutyric Acid Ethyl Ester

H25927.18 H25927.09
H25927.18EA 426 EUR
H25927.18 H25927.09
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 97%
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate

Purity: 97% CAS No.: 571-55-1 Molecular Formula: C9H11F3O4 Formula Weight: 240.18 Flash Point: 104°(220°F) Refractive Index: 1.4290 Boiling Point: 80-82°/1mm MDL No.: MFCD02677683

Formel: C₉H₁₁F₃O₄
Molecular Weight: 240.18 g/mol
Siedepunkt: 80…82 °C (1 mmHg)
Dichte: 1.235
Flammpunkt: 104 °C (220 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD02677683
CAS-Nummer: 571-55-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR