5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95%

Lieferant: Thermo Fisher Scientific

H25946.09 H25946.03
H25946.09EA 314 EUR
H25946.09 H25946.03
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95%
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin
Formel: C₄₈H₃₈N₄O₄
MDL: MFCD00012072
CAS-Nummer: 22112-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR