3-Nitroanthranilsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-3-nitrobenzoic acid
H26001.09
H26001.09EA
67
EUR
H26001.09
3-Nitroanthranilsäure 97%
3-Nitroanthranilsäure
Purity: 97%
CAS No.: 606-18-8
Molecular Formula: C7H6N2O4
Formula Weight: 182.14
Melting Point: 210-212°
MDL No.: MFCD00024261
Formel:
C₇H₆N₂O₄ Molecular Weight: 182,14 g/mol Schmelzpunkt: 210…212 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024261 CAS-Nummer: 606-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...