2-Benzyloxy-2-methylpropan-1-ol 95%

Lieferant: Thermo Scientific
Achtung

H26006.03
H26006.03EA 259 EUR
H26006.03
2-Benzyloxy-2-methylpropan-1-ol 95%
2-Benzyloxy-2-methylpropan-1-ol

Purity: 95% CAS No.: 91968-71-7 Molecular Formula: C11H16O2 Formula Weight: 180.12 Boiling Point: 80-85°/0.1mm MDL No.: MFCD08459314

Formel: C₁₁H₁₆O₂
Siedepunkt: 80…85 °C (0,1 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD08459314
CAS-Nummer: 91968-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR