3,4-Bis(trifluormethyl)pyridin 96%

Lieferant: Thermo Scientific
Achtung

H26008.03
H26008.03EA 113 EUR
H26008.03
3,4-Bis(trifluormethyl)pyridin 96%
3,4-Bis(trifluormethyl)pyridin

Purity: 96% CAS No.: 20857-46-9 Molecular Formula: C7H3F6N Formula Weight: 215.10 Refractive Index: 1.3920 Boiling Point: 42-44°/24mm MDL No.: MFCD08686956

Formel: C₇H₃F₆N
Molecular Weight: 215,1 g/mol
Siedepunkt: 42…44 °C (24 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD08686956
CAS-Nummer: 20857-46-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR