1,4-Bis(trimethylsilyl)tetrafluorobenzene 98%

Lieferant: Thermo Scientific
Achtung

H26009.03
H26009.03EA 236 EUR
H26009.03
1,4-Bis(trimethylsilyl)tetrafluorobenzene 98%
1,4-Bis(trimethylsilyl)tetrafluorobenzene

Purity: 99% CAS No.: 16956-91-5 Molecular Formula: C12H18F4Si2 Formula Weight: 294.44 Melting Point: 53-55° MDL No.: MFCD00093109

Formel: C₁₂H₁₈F₄Si₂
Molecular Weight: 294,44 g/mol
Schmelzpunkt: 53…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00093109
CAS-Nummer: 16956-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR