(4-Chlorphenyl)methansulfonsäurechlorid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Chloro-α-toluenesulphonyl chloride
H26017.03
H26017.06
H26017.03EA
68.1
EUR
H26017.03
H26017.06
(4-Chlorphenyl)methansulfonsäurechlorid ≥97%
(4-Chlorophenyl)methanesulphonyl chloride
Purity: 97%
CAS No.: 6966-45-6
Molecular Formula: C7H6Cl2O2S
Formula Weight: 225.10
Melting Point: 93-96°
UN No.: 3261
MDL No.: MFCD01631927
Formel:
C₇H₆Cl₂O₂S Molecular Weight: 225.10 g/mol Schmelzpunkt: 93…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631927 CAS-Nummer: 6966-45-6 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...