5-Chlor-2-methoxy-4-methylbenzolsulfonylchlorid ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
5-Chlor-2-methoxy-4-methylbenzolsulfochlorid, 5-Chlor-2-methoxy-4-methylbenzolsulfonsäurechlorid
H26020.06
H26020.06EA
125
EUR
H26020.06
5-Chlor-2-methoxy-4-methylbenzolsulfonylchlorid ≥97%
5-Chlor-2-methoxy-4-methylbenzolsulfonylchlorid
Purity: 97%
CAS No.: 889939-48-4
Molecular Formula: C8H8Cl2O3S
Formula Weight: 255.12
Melting Point: 126-128°
UN No.: 3261
MDL No.: MFCD08445709
Formel:
C₈H₈Cl₂O₃S Molecular Weight: 255,12 g/mol Schmelzpunkt: 126…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08445709 CAS-Nummer: 889939-48-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...