3-Chlorpicolinsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Chloropyridine-2-carboxylic acid
H26024.03
H26024.03EA
84
EUR
H26024.03
3-Chlorpicolinsäure ≥97%
3-Chlorpicolinsäure
Purity: 97%
CAS No.: 57266-69-0
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Melting Point: 131° dec.
MDL No.: MFCD01646325
Formel:
C₆H₄ClNO₂ Molecular Weight: 157,56 g/mol Schmelzpunkt: 131 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646325 CAS-Nummer: 57266-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...