1,1-Cyclohexandicarbonsäure 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Cyclohexane-1,1-dicarboxylic acid
H26025.03
H26025.03EA
75.3
EUR
H26025.03
1,1-Cyclohexandicarbonsäure 95%
1,1-Cyclohexanedicarboxylic acid
Purity: 95%
CAS No.: 1127-08-8
Molecular Formula: C8H12O4
Formula Weight: 172.18
Odour: Odourless
Melting Point: ca 175° dec.
MDL No.: MFCD00094668
Formel:
C₈H₁₂O₄ Molecular Weight: 172,18 g/mol Schmelzpunkt: ∼ 175 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00094668 CAS-Nummer: 1127-08-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...