4,4-Difluorcyclohexanon ≥97%

Lieferant: Thermo Scientific
Achtung

H26027.03
H26027.03EA 473 EUR
H26027.03
4,4-Difluorcyclohexanon ≥97%
4,4-Difluorocyclohexanone

Purity: 97% CAS No.: 22515-18-0 Molecular Formula: C6H8F2O Formula Weight: 134.12 Melting Point: 34-40° MDL No.: MFCD08146635

Formel: C₆H₈F₂O
Molecular Weight: 134,12 g/mol
Schmelzpunkt: 34…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08146635
CAS-Nummer: 22515-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR