5-Fluor-1H-indazol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H26033.03
H26033.03EA 95.4 EUR
H26033.03
5-Fluor-1H-indazol ≥98%
5-Fluoro-1H-indazole

Purity: 98% CAS No.: 348-26-5 Molecular Formula: C7H5FN2 Formula Weight: 136.13 Melting Point: 122-124° MDL No.: MFCD04972877

Formel: C₇H₅FN₂
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04972877
CAS-Nummer: 348-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR