1,4-Dioxaspiro[4.5]decan-8-ol ≥90%
Lieferant: Thermo Scientific
Synonyme:
4-Hydroxycyclohexan-1-one monoethylene ketal, 4-Hydroxycyclohexanone ethylene acetal
H26037.03
H26037.03EA
71.9
EUR
H26037.03
1,4-Dioxaspiro[4.5]decan-8-ol ≥90%
1,4-Dioxaspiro[4.5]decan-8-ol
Purity: 90+%
CAS No.: 22428-87-1
Molecular Formula: C8H14O3
Formula Weight: 158.19
Refractive Index: 1.4880
MDL No.: MFCD00067003
Formel:
C₈H₁₄O₃ Molecular Weight: 158,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067003 CAS-Nummer: 22428-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...