4-(2-Pyrrolidinyl)pyridin 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Pyrrolidin-2-ylpyridine

H26051.MD
H26051.MDEA 111 EUR
H26051.MD
4-(2-Pyrrolidinyl)pyridin 96%
4-(2-Pyrrolidinyl)pyridin

Purity: 96% CAS No.: 128562-25-4 Molecular Formula: C9H12N2 Formula Weight: 148.21 MDL No.: MFCD01862536

Formel: C₉H₁₂N₂
Molecular Weight: 148,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01862536
CAS-Nummer: 128562-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR