1-Thiophen-2-ylethylamine 90%, tech.
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-(Thiophen-2-yl)ethanamine, 2-(1-Aminoethyl)thiophene
H26052.ME
H26052.MEEA
688
EUR
H26052.ME
1-Thiophen-2-ylethylamine 90%, tech.
1-Thiophen-2-ylethylamine
Purity: 90%
CAS No.: 6309-16-6
Molecular Formula: C6H9NS
Formula Weight: 127.21
Refractive Index: 1.5475
UN No.: UN2735
MDL No.: MFCD02734311
Formel:
C₆H₉NS Molecular Weight: 127,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02734311 CAS-Nummer: 6309-16-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...