Triphenylsulphonium chloride 94%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H26059.03
H26059.03EA
85.6
EUR
H26059.03
Triphenylsulphonium chloride 94%
Triphenylsulphonium chloride
Purity: 94%
CAS No.: 4270-70-6
Molecular Formula: (C6H5)3SCl
Formula Weight: 298.84
MDL No.: MFCD00031716
Formel:
(C₆H₅)₃SCl Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031716 CAS-Nummer: 4270-70-6 EINECS: 224-259-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...