4-Cyanophenylacetic acid 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-(4-Cyanophenyl)acetic acid

H26062.03 H26062.06
H26062.03EA 130 EUR
H26062.03 H26062.06
4-Cyanophenylacetic acid 97%
4-Cyanophenylacetic acid

Purity: 97% CAS No.: 5462-71-5 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: 150-152° UN No.: UN3439 MDL No.: MFCD06798066

Formel: C₉H₇NO₂
Molecular Weight: 161.16 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06798066
CAS-Nummer: 5462-71-5
EINECS: 226-753-7
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR