3-Brom-5-fluorbenzonitril ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

H26066.09 H26066.03
H26066.09EA 137 EUR
H26066.09 H26066.03
3-Brom-5-fluorbenzonitril ≥98%
3-Brom-5-fluorbenzonitril

Purity: 98% CAS No.: 179898-34-1 Molecular Formula: C7H3BrFN Formula Weight: 200.01 UN No.: 3439 MDL No.: MFCD04038227

Formel: C₇H₃BrFN
Molecular Weight: 200.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038227
CAS-Nummer: 179898-34-1
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR