(4-Cyanphenyl)essigsäureethylester ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Ethyl-(4-cyanphenyl)acetat

H26078.03
H26078.03EA 107 EUR
H26078.03
(4-Cyanphenyl)essigsäureethylester ≥98%
(4-Cyanphenyl)essigsäureethylester

Purity: 98% CAS No.: 1528-41-2 Molecular Formula: C11H11NO2 Formula Weight: 189.21 Melting Point: 90-94° UN No.: 3439 MDL No.: MFCD09953133

Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,21 g/mol
Schmelzpunkt: 90…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09953133
CAS-Nummer: 1528-41-2
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR