HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat) ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate
H26082.03
H26082.06
H26082.14
H26082.03EA
36.5
EUR
H26082.03
H26082.06
H26082.14
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat) ≥99%
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)
Purity: 99%
CAS No.: 148893-10-1
Molecular Formula: C10H15F6N6OP
Formula Weight: 380.23
Melting Point: ca 185° dec.
MDL No.: MFCD00274639
Formel:
C₁₀H₁₅F₆N₆OP Schmelzpunkt: ∼ 185 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00274639 CAS-Nummer: 148893-10-1 UN: 1325 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...