2,3-Difluor-4-hydroxybenzoesäure ≥96%
Lieferant: Thermo Scientific
|
Achtung
|
H26087.03
H26087.03EA
95
EUR
H26087.03
2,3-Difluor-4-hydroxybenzoesäure ≥96%
2,3-Difluoro-4-hydroxybenzoic acid
Purity: 99%
CAS No.: 175968-39-5
Molecular Formula: C7H4F2O3
Formula Weight: 174.10
Melting Point: 205-210°
MDL No.: MFCD01321116
Formel:
C₇H₄F₂O₃ Schmelzpunkt: 205…210 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01321116 CAS-Nummer: 175968-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...