3,4-Difluorsalicylsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
H26088.03
H26088.03EA
104
EUR
H26088.03
3,4-Difluorsalicylsäure ≥98%
3,4-Difluorsalicylsäure
Purity: 98%
CAS No.: 189283-51-0
Molecular Formula: C7H4F2O3
Formula Weight: 174.10
Melting Point: 177-179°
MDL No.: MFCD03428564
Formel:
C₇H₄F₂O₃ Molecular Weight: 174,1 g/mol Schmelzpunkt: 177…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03428564 CAS-Nummer: 189283-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...