4-Fluor-o-anisaldehyd ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Fluor-2-anisaldehyd, 4-Fluor-2-methoxybenzaldehyd
H26091.03
H26091.03EA
96.9
EUR
H26091.03
4-Fluor-o-anisaldehyd ≥98%
4-Fluor-o-anisaldehyd
Purity: 98%
CAS No.: 450-83-9
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Melting Point: 60-62°
MDL No.: MFCD00143318
Formel:
C₈H₇FO₂ Molecular Weight: 154,14 g/mol Schmelzpunkt: 60…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143318 CAS-Nummer: 450-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...