4-Amino-3-fluorbenzoesäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-3-fluorobenzoic Acid

H26092.03
H26092.03EA 70.5 EUR
H26092.03
4-Amino-3-fluorbenzoesäure 98%
4-Amino-3-fluorobenzoic acid

Purity: 98% CAS No.: 455-87-8 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Melting Point: 214-216° MDL No.: MFCD01660374

Formel: C₇H₆FNO₂
Molecular Weight: 155,13 g/mol
Schmelzpunkt: 214…216 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01660374
CAS-Nummer: 455-87-8
EINECS: 207-250-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR