4-Hydroxy-3-(trifluormethyl)benzoesäure ≥98%

Lieferant: Thermo Scientific
Achtung

H26097.03
H26097.03EA 119 EUR
H26097.03
4-Hydroxy-3-(trifluormethyl)benzoesäure ≥98%
4-Hydroxy-3-(trifluoromethyl)benzoic acid

Purity: 98% CAS No.: 220239-68-9 Molecular Formula: C8H5F3O3 Formula Weight: 206.12 Melting Point: 144-147° MDL No.: MFCD01091008

Formel: C₈H₅F₃O₃
Molecular Weight: 206,12 g/mol
Schmelzpunkt: 144…147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01091008
CAS-Nummer: 220239-68-9
EINECS: None

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR