2-Chlor-5-hydroxybenzoesäure 99%
Lieferant: Thermo Scientific
|
Achtung
|
H26112.03
H26112.03EA
204
EUR
H26112.03
2-Chlor-5-hydroxybenzoesäure 99%
2-Chloro-5-hydroxybenzoic acid
Purity: 99%
CAS No.: 56961-30-9
Molecular Formula: C7H5ClO3
Formula Weight: 172.57
Melting Point: 178-180°
MDL No.: MFCD04038818
Formel:
C₇H₅ClO₃ Molecular Weight: 172,57 g/mol Schmelzpunkt: 178…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038818 CAS-Nummer: 56961-30-9 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...