3-Nitro-5-(trifluormethyl)phenol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Hydroxy-5-nitrobenzotrifluorid
H26113.03
H26113.03EA
107
EUR
H26113.03
3-Nitro-5-(trifluormethyl)phenol ≥98%
3-Nitro-5-(trifluormethyl)phenol
Purity: 98%
CAS No.: 349-57-5
Molecular Formula: C7H4F3NO3
Formula Weight: 207.11
Melting Point: 89-92°
MDL No.: MFCD04973779
Formel:
C₇H₄F₃NO₃ Molecular Weight: 207,11 g/mol Schmelzpunkt: 89…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973779 CAS-Nummer: 349-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...