5-Fluoro-2-nitrophenylacetonitrile 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-(5-Fluoro-2-nitrophenyl)acetonitrile
H26125.03
H26125.03EA
75.9
EUR
H26125.03
5-Fluoro-2-nitrophenylacetonitrile 99%
5-Fluoro-2-nitrophenylacetonitrile
Purity: 99%
CAS No.: 3456-75-5
Molecular Formula: C8H5FN2O2
Formula Weight: 180.14
Melting Point: 68-72°
UN No.: UN3439
MDL No.: MFCD00039742
Formel:
C₈H₅FN₂O₂ Molecular Weight: 180,14 g/mol Schmelzpunkt: 68…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039742 CAS-Nummer: 3456-75-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...