2-Methoxy-5-(trifluormethyl)benzaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H26135.03
H26135.03EA 173 EUR
H26135.03
2-Methoxy-5-(trifluormethyl)benzaldehyd ≥98%
2-Methoxy-5-(trifluoromethyl)benzaldehyde

Purity: 98% CAS No.: 146539-83-5 Molecular Formula: C9H7F3O2 Formula Weight: 204.15 Melting Point: 85-89° MDL No.: MFCD03788528

Formel: C₉H₇F₃O₂
Molecular Weight: 204.15 g/mol
Schmelzpunkt: 85…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788528
CAS-Nummer: 146539-83-5
EINECS: None

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR