2-Fluor-6-methoxybenzylbromid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-(Bromomethyl)-1-fluoro-3-methoxybenzene
H26136.03
H26136.MD
H26136.03EA
109
EUR
H26136.03
H26136.MD
2-Fluor-6-methoxybenzylbromid ≥98%
2-Fluor-6-methoxybenzylbromid
Purity: 98%
CAS No.: 500912-17-4
Molecular Formula: C8H8BrFO
Formula Weight: 219.05
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5570
Melting Point: 18-21°
UN No.: 3265
MDL No.: MFCD08688579
Formel:
C₈H₈BrFO Schmelzpunkt: 18…21 °C |
MDL:
MFCD08688579 CAS-Nummer: 500912-17-4 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...