4-Chlor-3-fluoranilin ≥99%

Lieferant: Thermo Scientific
Gefahr

H26148.03
H26148.03EA 82.5 EUR
H26148.03
4-Chlor-3-fluoranilin ≥99%
4-Chlor-3-fluoranilin

Purity: 99% CAS No.: 367-22-6 Molecular Formula: C6H5ClFN Formula Weight: 145.56 Odour: Characteristic Melting Point: 59-61° UN No.: 2811 MDL No.: MFCD01090987

Formel: C₆H₅ClFN
Molecular Weight: 145,56 g/mol
Schmelzpunkt: 59…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01090987
CAS-Nummer: 367-22-6
EINECS: 206-683-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR