4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

H26154.MD H26154.03
H26154.MDEA 78.5 EUR
H26154.MD H26154.03
4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid 97%
4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid

Purity: 97% CAS No.: 878572-17-9 Molecular Formula: C8H3F4NO4 Formula Weight: 253.11 Melting Point: 120-124° MDL No.: MFCD08282778

Formel: C₈H₃F₄NO₄
Schmelzpunkt: 120…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08282778
CAS-Nummer: 878572-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR